Please login to your online account to display your discounted pricing

Acetic Acid, Glacial, Aldehyde Free, for Cholesterol Determination, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$167.47 - $368.01

Chemical Identifiers

Chemical Name or Material Acetic Acid
CAS 64-19-7
Name Note Glacial, Aldehyde Free
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CH3COOH
Formula Weight 60.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002122

 
avantor performance
951105
Glass Bottle 2.5L Each for $368.01

02002133

 
avantor performance
951102
Glass Bottle 500mL Each for $167.47
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 60.05
Chemical Name or Material Acetic Acid
Molecular Formula CH3COOH
CAS 64-19-7
Quantity 2.5L
Flash Point 39.00°C
Name Note Glacial, Aldehyde Free
Packaging Glass Bottle
Specific Gravity 1L = 1.05kg