Please login to your online account to display your discounted pricing

Acetic Acid, Glacial ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Acetic Acid, Glacial
CAS 64-19-7
Purity Grade ACS Grade
Molecular Formula CH3COOH
Formula Weight 60.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK8817212

 
avantor performance
881746
SAFEMOR 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK8817500

 
avantor performance
881714
Glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.05
Purity Grade ACS Grade
Quantity 2.5L
Packaging SAFEMOR
Chemical Name or Material Acetic Acid, Glacial
Molecular Formula CH3COOH
CAS 64-19-7