missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic acid, glacial, ACS, 99.7+%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 36289AP

 View more versions of this product

Catalog No. AA36289AP



Description

Description

Specifications

Specifications

Acetic Acid
40°C (104°F)
118.1°C
500mL
64-19-7
C2H4O2
2789
acetasol, acetic acid, glacial, acide acetique, essigsaeure, ethanoic acid, ethylic acid, glacial, methanecarboxylic acid, vinegar acid
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
≥99.7%
1.049
1.3721
16.6°C
Glacial
99.7+%
MFCD00036152
14,55
Miscible with water.
CC(=O)O
60.052
CHEBI:15366
Liquid
ACS

Chemical Identifiers

64-19-7
60.052
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
acetasol, acetic acid, glacial, acide acetique, essigsaeure, ethanoic acid, ethylic acid, glacial, methanecarboxylic acid, vinegar acid
CHEBI:15366
CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226

P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL

EINECSNumber : 200-580-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents