Please login to your online account to display your discounted pricing

Acetic acid, glacial, 99+%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Acetic acid, glacial
CAS 64-19-7
Molecular Formula CH3CO2H
Formula Weight 60.05
MDL Number MFCD00036152
UN Number UN2789
Merck Index 14,55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA33252M4

  SDS
alfa aesar
33252M4
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA33252M6

  SDS
alfa aesar
33252M6
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.05
Quantity 20L
Chemical Name or Material Acetic acid, glacial
TSCA TSCA
Melting Point 16.6°C
pH 2.5
Boiling Point 118.1°C
Flash Point 40°C (104°F)
Physical Form Liquid
Assay ≥99%
Molecular Formula CH3CO2H
MDL Number MFCD00036152
CAS 64-19-7
UN Number UN2789
Merck Index 14,55
Density 1.049
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H226;
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Pungent
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
Refractive Index 1.3721
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H226;
Causes severe skin burns and eye damage.
Flammable liquid and vapour.