Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetic acid, Environmental Grade, 99% min

Chemical Identifiers

Chemical Name or Material Acetic acid
CAS 64-19-7
Purity Grade Environmental Grade
Formula Weight 60.05g/mol
MDL Number MFCD00036152
UN Number UN2789
Merck Index 14,55
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38740AP

 
alfa aesar™
38740-AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 1.049g/mL
Odor Pungent
Quantity 500mL
Refractive Index 1.3721
Melting Point 16.6°C
pH 2.5
Boiling Point 118.1°C
Quantity 500mL
Flash Point 40° (104°F)
Chemical Name or Material Acetic acid
CAS 64-19-7
Purity Grade Environmental Grade
Physical Form Liquid
Assay ≤99%
Formula Weight 60.05g/mol
MDL Number MFCD00036152
UN Number UN2789
Merck Index 14,55
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.