Please login to your online account to display your discounted pricing

Acetic Acid, Dilute R (120g/L), Ricca Chemical


Quantity 4L
Packaging Poly natural

Manufacturer: ricca chemical company  RI1361

 View more versions of this product

Catalog No. 136-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 120g/L
CAS 7732-18-5 (Water, Deionized)
Packaging Poly natural
CAS 64-19-7 (Acetic Acid)
Chemical Name or Material Acetic Acid, Dilute
Name Note Dilute Reagent

Specifications

Chemical Name or Material Acetic Acid, Dilute
Name Note Dilute Reagent
CAS 64-19-7 (Acetic Acid)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid