Please login to your online account to display your discounted pricing

Acetic Acid, Dilute R (120g/L), Ricca Chemical

$262.42 - $262.42

Chemical Identifiers

Chemical Name or Material Acetic Acid, Dilute
Name Note Dilute Reagent
CAS 64-19-7 (Acetic Acid)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

136-1

  SDS
ricca chemical company
RI1361
Poly natural 4L Each for $262.42

136-16

  SDS
ricca chemical company
RI13616
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R13632

 
ricca chemical company
RI13632
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 120g/L
CAS 7732-18-5 (Water, Deionized)
Packaging Poly natural
CAS 64-19-7 (Acetic Acid)
Chemical Name or Material Acetic Acid, Dilute
Name Note Dilute Reagent