Please login to your online account to display your discounted pricing

Acetic acid-D4, deuteration degree min. 99.5%, for NMR spectroscopy, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 10 x 0.75mL

Manufacturer: emd millipore corporation  8150360009

 View more versions of this product

Catalog No. M8150360009

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 64.08
Quantity 10 x 0.75mL
Color Colorless
Physical Form Liquid
Packaging Glass ampoule
Vapor Pressure 15.4 hPa (20°C)
Odor Stinging
Ignition Point 485°C
Molecular Formula C2D4O2 (Hill)
CAS 1186-52-3
Chemical Name or Material Acetic acid-D4
Assay Percent Range 99.5% min.

Specifications

Chemical Name or Material Acetic acid-D4
CAS 1186-52-3
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C2D4O2 (Hill)
Formula Weight 64.08