Please login to your online account to display your discounted pricing

Acetic acid-D4, deuteration degree min. 99.5%, for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$195.84 - $195.84

Chemical Identifiers

Chemical Name or Material Acetic acid-D4
CAS 1186-52-3
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C2D4O2 (Hill)
Formula Weight 64.08
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

M8150360010

 
emd millipore corporation
8150360010
10mL Each for $195.84
Description & Specifications

Specifications

Chemical Name or Material Acetic acid-D4
CAS 1186-52-3
Odor Stinging
Packaging Glass ampoule
Ignition Point 485°C
Vapor Pressure 15.4 hPa (20°C)
Physical Form Liquid
Quantity 10mL
Assay Percent Range 99.5% min.
Color Colorless
Molecular Formula C2D4O2 (Hill)
Formula Weight 64.08