Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetic acid-d4, 99.5% (Isotopic)

Chemical Identifiers

Chemical Name or Material Acetic acid-d4
CAS 1186-52-3
Name Note Isotopic
Formula Weight 64.08
MDL Number MFCD00051051
UN Number UN2789
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3646906

  SDS
alfa aesar™
3646906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3646909

  SDS
alfa aesar™
3646909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Acetic acid-d4
Name Note Isotopic
CAS 1186-52-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Odor Pungent, makes eyes water
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
Sensitivity Moisture Sensitive
Density 1.12g/mL
Boiling Point 118°C
Melting Point 17°C
Physical Form Liquid
Assay 99.5%
Quantity 5g
Formula Weight 64.08
MDL Number MFCD00051051
UN Number UN2789
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.