Please login to your online account to display your discounted pricing

Acetic Acid, Certified, 11.1% (v/v) ±0.3%, LabChem™

$73.80 - $73.80

Chemical Identifiers

Chemical Name or Material Acetic Acid
CAS 7732-18-5
CAS Max % 88.53
CAS 64-19-7
CAS Max % 11.47
Purity Grade Certified Grade
Linear Formula CH3COOH
Formula Weight 60.05
UN Number UN2790
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC102802

 
lab chem
LC102802
1L Each for $73.80
Description & Specifications

Specifications

Chemical Name or Material Acetic Acid
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 88.53
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS 64-19-7
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 11.47
Recommended Storage Room Temperature
Purity Grade Certified Grade
Physical Form Liquid
Assay 11.1% ± 0.3% (v/v)
Linear Formula CH3COOH
Formula Weight 60.05
UN Number UN2790
Density 1.01g/mL
Identification Passes Test
Odor Vinegar
Packaging Poly Bottle
Specific Gravity 1.01g/mL
Solubility Soluble in water
Traceability to NIST Traceable to NIST
Decomposition Information Carbon monoxide, Carbon dioxide, Corosive vapors
Color Clear, colorless
Quantity 1L
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.

Room Temperature