missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid, Certified, 0.100N ±0.001N (0.1M), LabChem™
Supplier: LabChem LC103502
Specifications
Acetic Acid | |
99.4,0.6 | |
C2H4O2 | |
MFCD00036152 | |
Soluble in water | |
CC(O)=O | |
0.100N ±0.001N (0.1M) | |
176 | |
60.05 | |
1g/mL | |
Vinegar | |
Liquid | |
Traceable to NIST |
64-19-7,7732-18-5 | |
Colorless | |
CH3COOH | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
60.05 | |
CHEBI:15366 | |
Certified | |
1 L | |
Poly Bottle | |
1g/mL |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature