Please login to your online account to display your discounted pricing

Acetic acid, ammonium salt-d7, 99 atom % D


Manufacturer: thermo fisher scientific  AC425860010

 View more versions of this product

Catalog No. AC425860010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 84.1
Quantity 1g
Linear Formula CD3CO2ND4
Chemical Name or Material Acetic acid, ammonium salt-d7
Molecular Formula C2D7NO2
CAS 194787-05-8
Packaging Glass bottle

Specifications

Chemical Name or Material Acetic acid, ammonium salt-d7
CAS 194787-05-8
Molecular Formula C2D7NO2
Linear Formula CD3CO2ND4
Formula Weight 84.1