missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic acid, ammonium salt-d7, 99 atom % D

Chemical Identifiers

CAS 194787-05-8
Molecular Formula C2D7NO2
Linear Formula CD3CO2ND4
Formula Weight 84.13
Chemical Name or Material Acetic acid, ammonium salt-d7
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
AC425860010 SDS Thermo Fisher Scientific
AC425860010
N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC425860050 SDS Thermo Fisher Scientific
AC425860050
N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

194787-05-8
CD3CO2ND4
Acetic acid, ammonium salt-d7
C2D7NO2
84.13

Specifications

194787-05-8
1g
CD3CO2ND4
Acetic acid, ammonium salt-d7
Glass bottle
C2D7NO2
84.1