missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic acid, ammonium salt, 98+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 218365000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysisSpecifications
Acetic acid, ammonium salt | |
Anhydrous | |
631-61-8 | |
100.0 | |
98+% | |
CH3CO2NH4 | |
02, 96 | |
15, 492 | |
Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C) | |
CC(=O)[O-].[NH4+] | |
77.083 | |
CHEBI:62947 | |
≥98% | |
500 g | |
Authentic | |
Crystals or Crystalline Powder |
6.5 to 7.5 (5% soln. at 20°C) | |
114.0°C | |
98.0 | |
White | |
C2H7NO2 | |
MFCD00013066 | |
01,38; 07,11 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
azanium;acetate | |
517165 | |
77.08 | |
Analytical | |
136°C | |
Plastic bottle |
Chemical Identifiers
631-61-8 | |
77.083 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
517165 | |
azanium;acetate |
C2H7NO2 | |
MFCD00013066 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CHEBI:62947 | |
CC(=O)[O-].[NH4+] |
RUO – Research Use Only