missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetic acid, Acculute Standard Volumetric Solution, Final Concentration 1.0N

Manufacturer:  Alfa Aesar™ 88556LQ

 View more versions of this product

Catalog No. AA88556LQ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Acetic acid Acculute Standard Volumetric Solution
64-19-7
CH3CO2H
UN2790
CC(=O)O
60.052
CHEBI:15366
Liquid
1 unit
Final Concentration 1.0N
C2H4O2
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
Pungent

Chemical Identifiers

64-19-7
60.052
QTBSBXVTEAMEQO-UHFFFAOYSA-N
CHEBI:15366
CC(=O)O
C2H4O2
MFCD00036152
176
acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ACETIC ACID SOLUTION

TSCA : TSCA

SDS Documents