Please login to your online account to display your discounted pricing

Acetic acid, 99+%, extra pure, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Acetic acid
CAS 64-19-7
Purity Grade Extra Pure
Assay Percent Range 99+%
Molecular Formula C2H4O2
Linear Formula CH3COOH
Formula Weight 60.05
Synonym Glacial acetic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC295310050

 
thermo fisher scientific
AC295310050
Plastic Jerrycan 5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC295310250

 
thermo fisher scientific
AC295310250
Metal drum 25L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Extra Pure
Formula Weight 60.05
Quantity 5L
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Packaging Plastic Jerrycan
Density 1.0480g/mL
Chemical Name or Material Acetic acid
Assay Percent Range 99+%
Molecular Formula C2H4O2
Linear Formula CH3COOH
CAS 64-19-7
Synonym Glacial acetic acid
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Safety and Handling

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.