Please login to your online account to display your discounted pricing

Acetic acid, 99.8%, for biochemistry, ACROS Organics™

$106.95 - $183.97

Chemical Identifiers

Chemical Name or Material Acetic Acid
Name Note For biochemistry
CAS 64-19-7
CAS Min % 99.75
CAS Max % 100.0
Purity Grade For biochemistry
Assay ≥99.7%
Assay Percent Range 99.8%
Molecular Formula C2H4O2
Linear Formula CH3COOH
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC148930100

  SDS
thermo fisher scientific
AC148930100
Plastic Jerrycan 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC148930010

  SDS
thermo fisher scientific
148930010
Glass Bottle 1L Each for $106.95

AC148930025

  Accessories SDS
thermo fisher scientific
148930025
Glass Bottle 2.5L Each for $183.97
Description & Specifications

Specifications

Assay ≥99.7%
Formula Weight 60.05
Physical Form Liquid
Quantity 10L
Color Clear
Beilstein 02, 96
Merck Index 15, 54
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Packaging Plastic Jerrycan
Density 1.0480g/mL
Purity Grade For biochemistry
Fieser 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12
CAS Max % 100.0
Sulfate 1ppm max.
Chloride 1ppm max.
Iron (Fe) 0.2ppm max.
Absorbance (neat)
Linear Formula CH3COOH
Additional Information Vapor Pressure: 1.52kPa at 20°C
Substances Reducing Dichromate Passes Test
Dilution Test Passes Test
Absorbance Conditions (neat)
Cadmium (Cd) 0.1ppm max.
Chemical Name or Material Acetic acid
Assay Percent Range 99.8%
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Name Note For biochemistry
CAS Min % 99.75
Substances Reducing Permanganate Passes Test
Synonym Glacial acetic acid
Heavy Metals (as Pb) 5ppm max.
Nickel (Ni) 0.1ppm max.
Substit Aromatic Aldehydes Passes Test
Molecular Formula C2H4O2
Infrared Spectrum Authentic
CAS 64-19-7
MDL Number MFCD00036152
Safety and Handling

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.