Please login to your online account to display your discounted pricing

Acetic acid, 99.8%, for analysis, ACROS Organics™


Quantity 5L
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC222140050

 View more versions of this product

Catalog No. AC222140050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥99.8%
Formula Weight 60.05
Quantity 5L
Color Clear
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Packaging Plastic Jerrycan
Purity Grade For analysis
Density 1.0480g/mL
Beilstein 02, 96
Fieser 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12
Merck Index 15, 54
CAS Max % 100.0
Substances Reducing Permanganate Passes Test
Substances Reducing Dichromate Passes Test
Water 0.2% max.
Linear Formula CH3COOH
Synonym Glacial acetic acid
Copper (Cu) 0.1ppm max.
Dilution Test Passes Test
Nickel (Ni) 0.1ppm max.
Chemical Name or Material Acetic acid
Molecular Formula C2H4O2
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Sulfate 1ppm max.
Name Note For Analysis
Assay Percent Range 99.8%
MDL Number MFCD00036152
CAS 64-19-7
CAS Min % 99.75
Heavy Metals (as Pb) 0.5ppm max.
Residue after Evaporation 0.001% max.
Arsenic (As) 0.5ppm max.
Cadmium (Cd) 0.1ppm max.
Acetic Anhydride 0.01% max.
Iron (Fe) 0.2ppm max.
Chlorine (Cl) 1ppm max.

Specifications

Chemical Name or Material Acetic acid
Name Note For Analysis
CAS 64-19-7
CAS Min % 99.75
CAS Max % 100.0
Purity Grade For analysis
Assay ≥99.8%
Assay Percent Range 99.8%
Molecular Formula C2H4O2
Linear Formula CH3COOH
Formula Weight 60.05
MDL Number MFCD00036152
Synonym Glacial acetic acid

For analysis

Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents