missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic acid, 99.7+%, ACS reagent, ACROS Organics™
$69.21 - $175.87
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.052 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | acetasol, acetic acid, glacial, acide acetique, essigsaeure, ethanoic acid, ethylic acid, glacial, methanecarboxylic acid, vinegar acid |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC423225000
|
Thermo Fisher Scientific
423225000 |
500mL | Glass Bottle |
Each for $69.21
|
|
AC423220025
|
Thermo Fisher Scientific
423220025 |
2.5L | Glass Bottle |
Each for $175.87
|
|
AC423220100
|
Thermo Fisher Scientific
AC423220100 |
10L | Plastic drum |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
64-19-7 | |
60.052 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
acetasol, acetic acid, glacial, acide acetique, essigsaeure, ethanoic acid, ethylic acid, glacial, methanecarboxylic acid, vinegar acid | |
CHEBI:15366 | |
CC(=O)O |
Specifications
64-19-7 | |
99.7 | |
Vapor Pressure: 1.52kPa at 20°C | |
C2H4O2 | |
1ppm max. | |
02, 96 | |
15, 54 | |
1.0480g/mL | |
CC(=O)O | |
Passes Test | |
176 | |
60.05 | |
≥99.70% | |
0.5ppm max. | |
Glass Bottle | |
Passes Test | |
1ppm max. | |
Undesignated | |
Acetic Acid |
0.01% max. | |
100.0 | |
99.7+% | |
CH3COOH | |
MFCD00036152 | |
02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12 | |
acetasol, acetic acid, glacial, acide acetique, essigsaeure, ethanoic acid, ethylic acid, glacial, methanecarboxylic acid, vinegar acid | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
60.052 | |
CHEBI:15366 | |
Liquid | |
ACS Reagent | |
0.2ppm max. | |
0.001% max. | |
Passes Test | |
0.0004 meq/g max. | |
500mL |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger