Please login to your online account to display your discounted pricing

Acetic Acid, 80% w/w (Certified), Fisher Chemical

$222.07 - $494.93

Chemical Identifiers

Chemical Name or Material Acetic Acid
Name Note 80% (w/w)
CAS 64-19-7
CAS 7732-18-5
Purity Grade Certified Grade
Assay 80 to 82%
Assay Percent Range 80 to 82 %
Molecular Formula C2H4O2
Linear Formula CH3COOH
Formula Weight 60.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A37-4

  SDS
fisher scientific company
A374
Glass Bottle 4L Each for $494.93
Case of 4 Each for $1,979.72

A37-1

  SDS
fisher scientific company
FLA371
Glass Bottle 1L Each for $222.07
Case of 6 Each for $1,332.42
Description & Specifications

Specifications

Melting Point −7°C
Purity Grade Certified Grade
Boiling Point 117°C
Formula Weight 60.05
Quantity 4L
DOT Information DOT Class 8, : Corrosive
Assay 80 to 82%
Physical Form Liquid
Normality 14.2
Color Colorless
pH 2.4
Vapor Pressure 16mmHg at 25°C
Sulfate 4ppm max.
Assay Percent Range 80 to 82 %
Residue after Evaporation 0.002% max.
Substances Reducing Permanganate Pass Test
Iron (Fe) 4ppm max.
Health Hazard 2 DANGER!
Packaging Glass Bottle
Health Hazard 3 Emergency Overview
Combustible liquid. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:2
Instability:0
Cleanup Symbol Acid
ChemAlert Storage Symbol White
MDL Number MFCD00036152
Chloride 4ppm max.
CAS 7732-18-5
Heavy Metals (as Pb) 4ppm max.
Identification Pass Test
Additional Information Vapor Pressure: 16mmHg at 25°C
Formic or Sulfurous Acid Pass Test
CAS 64-19-7
Chemical Name or Material Acetic Acid
Linear Formula CH3COOH
Name Note 80% (w/w)
Molecular Formula C2H4O2

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Safety and Handling
 

Emergency Overview
Combustible liquid. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:2
Instability:0

DANGER!