Please login to your online account to display your discounted pricing

Acetic Acid, 5% v/v (For AOAC Method 953.08), Fisher Chemical

$43.79 - $137.73

Chemical Identifiers

Chemical Name or Material Acetic Acid, 5% v/v
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 96.0
CAS 64-19-7
CAS Min % 4.0
CAS Max % 6.0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SA9050-500

 
ricca chemical company
SA9050500
Poly Bottle 500mL Each for $43.79

SA9050-4

 
ricca chemical company
SA90504
Poly Bottle 4L Each for $137.73

SA9050-1

 
ricca chemical company
SA90501
Poly Bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Methods For AOAC Method 953.08
NIST Traceable to NIST.
ChemAlert Storage Symbol Gray
CAS Min % 94.0
Packaging Poly Bottle
Chemical Name or Material Acetic Acid, 5% v/v
CAS 7732-18-5
CAS Max % 96.0
CAS 64-19-7
CAS Min % 4.0
CAS Max % 6.0
Safety and Handling