Please login to your online account to display your discounted pricing

Acetic Acid, 5% (v/v) Aqueous Solution, Ricca Chemical


Quantity 20L
Packaging Cubitainer™

Manufacturer: ricca chemical company  RI1305

 View more versions of this product

Catalog No. 1305

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Clear colorless
Physical Form Liquid
Quantity 20L
CAS Max % 96.0
Chemical Name or Material Acetic Acid
Solvent or Matrix Aqueous Solution
Concentration or Composition Notes 5% (v/v)
Name Note 5% (v/v)
CAS 7732-18-5
CAS Min % 94.0
CAS 64-19-7
CAS Min % 4.0
Packaging Cubitainer™
CAS Max % 6.0

Specifications

Chemical Name or Material Acetic Acid
Name Note 5% (v/v)
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 96.0
CAS 64-19-7
CAS Min % 4.0
CAS Max % 6.0
Physical Form Liquid