Please login to your online account to display your discounted pricing

Acetic Acid, 5% (v/v) Aqueous Solution, Ricca Chemical


Quantity 4L
Packaging Poly Bottle

Manufacturer: ricca chemical company  RI1301

 View more versions of this product

Catalog No. 130-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 5% (v/v)
Solvent or Matrix Aqueous Solution
Chemical Name or Material Acetic Acid
Packaging Poly Bottle
CAS 64-19-7
CAS Max % 96.0
CAS Min % 4.0
CAS Min % 94.0
Name Note 5% (v/v)
CAS Max % 6.0
CAS 7732-18-5

Specifications

Chemical Name or Material Acetic Acid
Name Note 5% (v/v)
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 96.0
CAS 64-19-7
CAS Min % 4.0
CAS Max % 6.0
Physical Form Liquid