Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetic acid, 4% v/v aq. soln.

Chemical Identifiers

Chemical Name or Material Acetic acid
Formula Weight 60.05
MDL Number MFCD00036152
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35572AP

  SDS
alfa aesar™
35572AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35572K2

  SDS
alfa aesar™
35572K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.05
Chemical Name or Material Acetic acid
TSCA TSCA
Quantity 500mL
Physical Form Liquid
MDL Number MFCD00036152
Concentration 4% v/v aq. soln.
Odor Pungent