missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic acid, 4% v/v aq. soln., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 035572AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetic acid | |
C2H4O2 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
acetic acid | |
176 | |
60.05 | |
Pungent |
64-19-7 | |
MFCD00036152 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
4% v/v aq. soln. | |
60.05 | |
CHEBI:15366 | |
500 mL | |
Liquid |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling
EINECSNumber : 200-580-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only