Please login to your online account to display your discounted pricing

Acetic Acid, 25% v/v (For AOAC Method 948.10), Fisher Chemical

$68.42 - $68.42

Chemical Identifiers

Chemical Name or Material Acetic Acid, 25% v/v
CAS 7732-18-5
CAS Min % 74.0
CAS Max % 76.0
CAS 64-19-7
CAS Min % 24.0
CAS Max % 26.0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SA9025-1

  SDS
ricca chemical company
SA90251
Poly Bottle 1L Each for $68.42
Description & Specifications

Specifications

Quantity 1L
Methods For AOAC Method 948.10
NIST Traceable to NIST.
Chemical Name or Material Acetic Acid, 25% v/v
CAS 7732-18-5
ChemAlert Storage Symbol White
CAS Min % 74.0
CAS Max % 76.0
CAS 64-19-7
CAS Min % 24.0
Packaging Poly Bottle
CAS Max % 26.0
Safety and Handling