Please login to your online account to display your discounted pricing

Acetic Acid, 2% (v/v) Aqueous Solution, Ricca Chemical


Quantity 500mL
Packaging Poly bottle

Manufacturer: ricca chemical company  RI11016

 View more versions of this product

Catalog No. 110-16

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Acetic Acid
CAS 64-19-7 (Acetic Acid)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid
Packaging Poly bottle
Solvent or Matrix Aqueous Solution
Concentration or Composition Notes 2% (v/v)
Color Clear colorless
Quantity 500mL

Specifications

Chemical Name or Material Acetic Acid
CAS 64-19-7 (Acetic Acid)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid