missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid, 1.00 N, Ricca Chemical
Shelf Life: 18 months
Supplier: Ricca Chemical Company RI15025
Specifications
Acetic Acid 1N | |
92.0,5.56 | |
C2H4O2 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
10 L |
64-19-7,7732-18-5 | |
96.0,5.79 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(=O)O | |
60.052 | |
CHEBI:15366 | |
Cubitainer |