missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Acid 1L, Ricca Chemical

Shelf Life: 18 months

Manufacturer:  Ricca Chemical Company RI1505

 View more versions of this product

Catalog No. 150-5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Acetic Acid 1N
92.0,5.56
C2H4O2
CC(=O)O
60.052
CHEBI:15366
20L
64-19-7,7732-18-5
96.0,5.79
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
Cubitainer

Chemical Identifiers

64-19-7
60.052
176
C2H4O2
QTBSBXVTEAMEQO-UHFFFAOYSA-N
CHEBI:15366
SDS Documents