missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Acid 1L, Ricca Chemical

Shelf Life: 18 months

Manufacturer:  Ricca Chemical Company RI15032

 View more versions of this product

Catalog No. 150-32

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Acetic Acid 1N
92.0,5.56
C2H4O2
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
Poly Bottle
64-19-7,7732-18-5
96.0,5.79
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, glacial, acetasol, acide acetique, essigsaeure
CC(=O)O
60.052
CHEBI:15366
1L

Chemical Identifiers

64-19-7
60.052
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(=O)O
C2H4O2
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
SDS Documents