Learn More
Acethydrazide, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 102351000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acethydrazide | |
1068-57-1 | |
100.0 | |
White | |
>110°C | |
Authentic | |
Glass bottle | |
CH3CONHNH2 | |
100 g | |
acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine | |
OFLXLNCGODUUOT-UHFFFAOYSA-N | |
acetohydrazide | |
14039 | |
74.08 | |
Moist Crystals and Chunks |
95% | |
94.0 | |
57.0°C to 60.0°C | |
129.0°C (18.0 mmHg) | |
Balance (50°C, 3 Hrs) | |
94% min. (on dry substance) | |
C2H6N2O | |
MFCD00007610 | |
02,191 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,chloroform | |
CC(=O)NN | |
74.08 | |
CHEBI:48978 | |
95% |
Chemical Identifiers
1068-57-1 | |
74.08 | |
OFLXLNCGODUUOT-UHFFFAOYSA-N | |
14039 | |
acetohydrazide |
C2H6N2O | |
MFCD00007610 | |
acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine | |
CHEBI:48978 | |
CC(=O)NN |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
Flammable solid.
Risk of explosion if heated under confinement.
GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 213-948-7
RTECSNumber : AI1225000
TSCA : TSCA
RUO – Research Use Only