missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acethydrazide, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC102355000

 View more versions of this product

Catalog No. AC102355000



Specifications

Specifications

Acethydrazide
Authentic
Glass bottle
White
500g
1068-57-1
100.0
C2H6N2O
MFCD00007610
OFLXLNCGODUUOT-UHFFFAOYSA-N
acetohydrazide
14039
74.08
95%
>110°C
Balance (50°C, 3 Hrs)
129.0°C (18.0 mmHg)
57.0°C to 60.0°C
95%
94.0
95%
CH3CONHNH2
acethydrazide
CC(=O)NN
74.083
CHEBI:48978
Moist Crystals and Chunks

Chemical Identifiers

1068-57-1
74.083
OFLXLNCGODUUOT-UHFFFAOYSA-N
14039
acetohydrazide
C2H6N2O
MFCD00007610
acethydrazide
CHEBI:48978
CC(=O)NN
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
Flammable solid.
Risk of explosion if heated under confinement.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 213-948-7

RTECSNumber : AI1225000

TSCA : TSCA

SDS Documents