missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetate Buffer TS, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,631-61-8,64-19-7
CAS Min % 74.0,23.68,0.39
CAS Max % 77.0,24.64,0.4
Physical Form Liquid
Chemical Name or Material Acetate Buffer, Test Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
62-16 Ricca Chemical Company
RI6216
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
62-32 SDS Ricca Chemical Company
RI6232
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,631-61-8,64-19-7
77.0,24.64,0.4
Acetate Buffer, Test Solution
74.0,23.68,0.39
Liquid

Specifications

Liquid
500mL
74.0,0.39,23.68
77.0,24.64,0.4
Clear colorless
7732-18-5,631-61-8,64-19-7
Poly natural
Acetate Buffer, Test Solution