Please login to your online account to display your discounted pricing

Acetate Buffer, Purified, for Zinc Analysis (Dithizone Method), Ricca Chemical

For Analysis of Zinc (Dithizone Method)

$202.30 - $202.30

Chemical Identifiers

Chemical Name or Material Acetate Buffer
CAS 7732-18-5
CAS Min % 81.0
CAS Max % 84.0
CAS 6131-90-4
CAS Min % 11.67
CAS Max % 12.14
CAS 64-19-7
CAS Min % 5.67
CAS Max % 5.9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

70-16

 
ricca chemical company
RI7016
Poly natural 500mL Each for $202.30

70-32

 
ricca chemical company
RI7032
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 500mL
Treatment(s) Purified
Chemical Name or Material Acetate Buffer
CAS Max % 5.9
CAS 7732-18-5
CAS Min % 81.0
CAS Max % 84.0
CAS 6131-90-4
Packaging Poly natural
CAS Min % 11.67
CAS Max % 12.14
CAS 64-19-7
CAS Min % 5.67