Please login to your online account to display your discounted pricing

Acetate Buffer pH 6.0, For Aluminum, Certified, LabChem™

$82.17 - $82.17

Chemical Identifiers

Chemical Name or Material Acetate Buffer pH 6.0
CAS 7732-18-5
CAS Max % 86.17
CAS 6131-90-4
CAS Max % 13.6
CAS 64-19-7
CAS Max % 0.23
Name Note For Aluminum
Purity Grade Certified Grade
Linear Formula CH3COOHNa·3H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC100701

 
lab chem
LC100701
500mL Each for $82.17
Description & Specifications

Specifications

Purity Grade Certified Grade
pH 6.00 ± 0.05
Formula Weight 136.08
Quantity 500mL
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 64-19-7
Color Clear, colorless
CAS Max % 86.17
Packaging Poly Bottle
Solubility Soluble in water
CAS Max % 0.23
Identification Passes Test
Traceability to NIST Traceable to NIST
CAS 7732-18-5
CAS 6131-90-4
CAS Max % 13.6
Name Note For Aluminum
Physical Form Liquid
Chemical Name or Material Acetate Buffer pH 6.0
Recommended Storage Room Temperature
Linear Formula CH3COOHNa·3H2O
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Decomposition Information Carbon monoxide, Carbon dioxide
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature