Please login to your online account to display your discounted pricing

Acetate Buffer pH 5.5, For Arsenic, Certified, LabChem™

$62.09 - $62.09

Chemical Identifiers

Chemical Name or Material Acetate Buffer pH 5.5
CAS 7732-18-5
CAS Max % 97.49
CAS 6131-90-4
CAS Max % 2.34
CAS 64-19-7
CAS Max % 0.17
Name Note For Arsenic
Purity Grade Certified Grade
Linear Formula CH3COOHNa·3H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC100052

 
lab chem
LC100052
1L Each for $62.09
Description & Specifications

Specifications

pH 5.50 ± 0.05
Purity Grade Certified Grade
Formula Weight 136.08
Chemical Name or Material Acetate Buffer pH 5.5
Decomposition Information Carbon monoxide, Carbon dioxide
CAS Max % 0.17
Color Clear, colorless
Quantity 1L
Name Note For Arsenic
Physical Form Liquid
Linear Formula CH3COOHNa·3H2O
CAS 7732-18-5
Identification Passes Test
CAS Max % 97.49
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 6131-90-4
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 2.34
Recommended Storage Room Temperature
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS 64-19-7
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature