Please login to your online account to display your discounted pricing

Acetate Buffer, pH 5.5, APHA, For Arsenic, Ricca Chemical

$79.97 - $86.67

Chemical Identifiers

CAS 7732-18-5
Physical Form Liquid
Assay APHA for Arsenic
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

6316

 
ricca chemical company
RI6316
500mL Each for $79.97

6332

 
ricca chemical company
RI6332
1L Each for $86.67
Description & Specifications

Specifications

Quantity 500mL
Assay APHA for Arsenic
Physical Form Liquid
Solubility Solubility in Water: Infinite
CAS 7732-18-5
Odor Acetic acid
Appearance Clear colorless liquid
pH Range 4 to 6
Recommended Storage General
Safety and Handling

General