missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetate Buffer, pH 4.6, for Bromide Analysis, Ricca Chemical

For Analysis of Bromide

$61.76 - $61.76

Chemical Identifiers

CAS 7732-18-5,7647-14-5,6131-90-4,64-19-7
CAS Min % 82.0,7.42,5.61,2.59
CAS Max % 86.0,7.72,5.83,2.7
Physical Form Liquid
Chemical Name or Material Acetate Buffer, pH 4.6
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
5416 Ricca Chemical Company
RI5416
Poly natural 500mL Each for $61.76
541 Ricca Chemical Company
RI541
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
54-32 Ricca Chemical Company
RI5432
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7647-14-5,6131-90-4,64-19-7
86.0,7.72,5.83,2.7
Acetate Buffer, pH 4.6
82.0,7.42,5.61,2.59
Liquid

Specifications

7732-18-5,7647-14-5,6131-90-4,64-19-7
86.0,7.72,5.83,2.7
Poly natural
500mL
82.0,7.42,5.61,2.59
Liquid
Clear colorless
Acetate Buffer, pH 4.6