Please login to your online account to display your discounted pricing

Acetate Buffer pH 4.6 to 4.7, For Bromide, Certified, LabChem™

$92.57 - $92.57

Chemical Identifiers

Chemical Name or Material Acetate Buffer pH 4.6 to 4.7
CAS 7732-18-5
CAS Max % 80.7
CAS 7647-14-5
CAS Max % 9
CAS 6131-90-4
CAS Max % 6.8
CAS 64-19-7
CAS Max % 3
CAS 1310-73-2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC100102

 
lab chem
LC100102
1L Each for $92.57
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 136.08
Chemical Name or Material Acetate Buffer pH 4.6 to 4.7
Decomposition Information Carbon monoxide, Carbon dioxide
CAS Max % 6.8
pH 4.6 to 4.7
Color Clear, colorless
Quantity 1L
CAS 64-19-7
CAS Max % 3
CAS 1310-73-2
CAS Max % 0.5
Name Note For Bromide
Physical Form Liquid
Linear Formula CH3COOHNa·3H2O
CAS 7732-18-5
Health Hazard 1 Warning
Identification Passes Test
CAS Max % 80.7
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
CAS 7647-14-5
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Packaging Poly Bottle
CAS Max % 9
Recommended Storage Room Temperature
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS 6131-90-4
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

Room Temperature