Please login to your online account to display your discounted pricing

Acetate Buffer pH 4.5, For Cyanide, Certified, LabChem™

$48.77 - $48.77

Chemical Identifiers

Chemical Name or Material Acetate Buffer pH 4.5
CAS 64-19-7
CAS Max % 36.9
CAS 6131-90-4
CAS Max % 33.6
CAS 7732-18-5
CAS Max % 29.5
Name Note For Cyanide
Purity Grade Certified Grade
Linear Formula CH3COOHNa·3H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC100401

 
lab chem
LC100401
500mL Each for $48.77
Description & Specifications

Specifications

pH 4.50 ± 0.05
Purity Grade Certified Grade
Formula Weight 136.08
Chemical Name or Material Acetate Buffer pH 4.5
Decomposition Information Carbon monoxide, Carbon dioxide
CAS Max % 29.5
Color Clear, colorless
Quantity 500mL
Name Note For Cyanide
Physical Form Liquid
Linear Formula CH3COOHNa·3H2O
CAS 64-19-7
Health Hazard 1 Warning
Identification Passes Test
CAS Max % 36.9
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
CAS 6131-90-4
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Packaging Poly Bottle
CAS Max % 33.6
Recommended Storage Room Temperature
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS 7732-18-5
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

Room Temperature