missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetate Buffer, pH 4.5, for Cyanide Determination, Ricca Chemical
Cyanide Determination
$92.66 - $92.66
Specifications
Chemical Name or Material | Acetate Buffer |
---|---|
Name Note | pH 4.5 |
CAS | 7732-18-5,64-19-7,6131-90-4 |
CAS Min % | 51.0,26.59,20.78 |
CAS Max % | 53.0,27.67,21.62 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
52-16
|
Ricca Chemical Company
RI5216 |
500 mL | Natural Poly Bottle |
Each for $92.66
|
|
||||
Specifications
Acetate Buffer | |
7732-18-5,64-19-7,6131-90-4 | |
53.0,27.67,21.62 | |
Liquid |
pH 4.5 | |
51.0,26.59,20.78 | |
Colorless | |
Natural Poly Bottle |