Acetate Buffer pH 4.0, For Chlorine, Iodine, Certified, LabChem™
Manufacturer:  LabChem LC100304
 View more versions of this product
Catalog No. LC100304
Specifications
Specifications
Acetate Buffer
64-19-7,7732-18-5,6131-90-4
CH3COOHNa·3H2O
Liquid
4.0 ±0.1
For Chlorine, Iodine
45.5,32.5,22
136.08
Certified
4L