Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetate, 1M buffer soln., pH, 4.0, low endotoxin


Quantity 100mL

Manufacturer: alfa aesar™  J67486AE

 View more versions of this product

Catalog No. AAJ67486AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Acetate
CAS 127-09-3
MDL Number MFCD00012459
Physical Form Liquid
pH 4.0
Quantity 100mL
Concentration 1M buffer soln.
TSCA TSCA
Name Note low endotoxin
Color Colorless

Specifications

Chemical Name or Material Acetate
CAS 127-09-3
Name Note low endotoxin
MDL Number MFCD00012459