Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetate, 1M buffer soln., pH, 4.0, low endotoxin

Chemical Identifiers

Chemical Name or Material Acetate
CAS 127-09-3
Name Note low endotoxin
MDL Number MFCD00012459
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67486AE

 
alfa aesar™
J67486AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67486AK

 
alfa aesar™
J67486AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Acetate
CAS 127-09-3
MDL Number MFCD00012459
Physical Form Liquid
pH 4.0
Quantity 100mL
Concentration 1M buffer soln.
TSCA TSCA
Name Note low endotoxin
Color Colorless