Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetate, 1M buffer soln., pH, 4.0, low endotoxin


Quantity 250mL

Manufacturer: alfa aesar™  J67486AK

 View more versions of this product

Catalog No. AAJ67486AK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Acetate
Physical Form Liquid
MDL Number MFCD00012459
pH 4.0
Color Colorless
Quantity 250mL
TSCA TSCA
Name Note low endotoxin
Concentration 1M buffer soln.
CAS 127-09-3

Specifications

Chemical Name or Material Acetate
CAS 127-09-3
Name Note low endotoxin
MDL Number MFCD00012459