missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Acetamidocinnamic acid, 98%, ACROS Organics™

$154.14 - $154.14

Chemical Identifiers

CAS 5469-45-4
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
InChI Key XODAOBAZOQSFDS-YFHOEESVSA-N
Synonym .alpha.-acetamidocinnamic acid
PubChem CID 5370579
IUPAC Name (Z)-2-acetamido-3-phenylprop-2-enoic acid
SMILES CC(=O)NC(=CC1=CC=CC=C1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC200261000 SDS View Documents Thermo Fisher Scientific
AC200261000
100g Glass bottle Each for $154.14
 
Specifications

Chemical Identifiers

5469-45-4
205.21
.alpha.-acetamidocinnamic acid
(Z)-2-acetamido-3-phenylprop-2-enoic acid
C11H11NO3
XODAOBAZOQSFDS-YFHOEESVSA-N
5370579
CC(=O)NC(=CC1=CC=CC=C1)C(=O)O

Specifications

Glass bottle
100g
97.5% min. (HPLC)
C6H5CH=C(NHCOCH3)CO2H
.alpha.-acetamidocinnamic acid
CC(=O)NC(=CC1=CC=CC=C1)C(=O)O
205.21
205.21
α-Acetamidocinnamic acid
188.0°C to 190.0°C
5469-45-4
C11H11NO3
10,683
XODAOBAZOQSFDS-YFHOEESVSA-N
(Z)-2-acetamido-3-phenylprop-2-enoic acid
5370579
98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 226-795-6