missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetamidine hydrochloride, 98+%, ACROS Organics™

Chemical Identifiers

CAS 124-42-5
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C2H6N2·ClH
Linear Formula CH3C(=NH)NH2·HCl
Formula Weight 94.54
MDL Number MFCD00013016
Chemical Name or Material Acetamidine hydrochloride
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC170215000 SDS Thermo Fisher Scientific
AC170215000
Glass bottle 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC170210100 SDS Fisher Scientific Acros
170210100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC170211000 SDS Fisher Scientific Acros
170211000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

124-42-5
100.0
C2H6N2·ClH
94.54
Acetamidine hydrochloride
98.0
98+%
CH3C(=NH)NH2·HCl
MFCD00013016
≥98.0%

Specifications

94.54
500g
160° to 170°C
15, 45
98.0
124-42-5
98+%
CH3C(=NH)NH2·HCl
Acetamidine hydrochloride
Crystalline Powder, Crystals and/or Chunks
White to cream
02, 185
100.0
Authentic
C2H6N2·ClH
MFCD00013016
Glass bottle
≥98.0%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning