Learn More
Acetamidine hydrochloride, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 170210100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Acetamidine hydrochloride | |
98.0 | |
Authentic | |
Glass bottle | |
CH3C(=NH)NH2·HCl | |
10g | |
15, 45 | |
Solubility in water: 1g/mL. Other solubilities: soluble in alcohol,insoluble in acetone | |
CC(=N)N.Cl | |
94.54 | |
94.54 |
124-42-5 | |
100.0 | |
98% min. (Argentometry) | |
C2H6N2·ClH | |
MFCD00013016 | |
02, 185 | |
acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride | |
WCQOBLXWLRDEQA-UHFFFAOYSA-N | |
ethanimidamide;hydrochloride | |
67170 | |
98+% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 204-700-9
RUO â Research Use Only