missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetamide, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 153635000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetamide | |
79.0°C to 81.0°C | |
221.0°C | |
98.5% min. (GC) | |
C2H5NO | |
500 g | |
01,3 | |
ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van | |
DLFVBJFMPXGRIB-UHFFFAOYSA-N | |
acetamide | |
178 | |
59.06 | |
Pure |
60-35-5 | |
White | |
Authentic | |
Plastic bottle | |
CH3CONH2 | |
02,II,177 | |
15,44 | |
Solubility in water: 2000g/L (20°C). Other solubilities: 1g/2 mL alcohol- 1g/6 mL pyridine,soluble in chloroform,glycerol and hot benzene,slightly soluble in ether | |
CC(=O)N | |
59.06 | |
CHEBI:49028 | |
99% | |
Crystals |
Chemical Identifiers
60-35-5 | |
59.06 | |
ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van | |
CHEBI:49028 | |
CC(=O)N |
C2H5NO | |
DLFVBJFMPXGRIB-UHFFFAOYSA-N | |
178 | |
acetamide |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 200-473-5
RUO – Research Use Only